siRNA Details
Gene Symbol: Ccdc30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270435.1 | NP_001257364.1 | 6 | 1039 | ||
NM_001270446.1 | NP_001257375.1 | 1 | 295 | |||
NM_028506.1 | NP_082782.1 | 2 | 302 | |||
NM_029286.2 | NP_083562.1 | 6 | 1054 | |||
GenBank mRNA | AK006668.1 | - | 4 | 600 | ||
AK043853.1 | - | 1 | 288 | |||
AK049010.1 | - | 1 | 195 | |||
AK078362.1 | - | 6 | 981 | |||
AK163185.1 | - | 6 | 980 | |||
AK224940.1 | - | 2 | 231 | |||
AK224959.1 | - | 1 | 203 | |||
AK224961.1 | - | 1 | 296 | |||
BC114436.1 | - | 6 | 981 | 40059415 |
