siRNA Details
Gene Symbol: Ttc21b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047604.2 | NP_001041069.1 | 3 | 261 | ||
NM_001290669.1 | NP_001277598.1 | 3 | 261 | |||
GenBank mRNA | BC151199.1 | - | 3 | 236 | ||
BC172115.1 | - | 3 | 236 | |||
DQ011164.1 | - | 3 | 261 |
