siRNA Details
Gene Symbol: Fam122c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028671.2 | NP_082947.1 | 2 | 157 | ||
GenBank mRNA | AK015288.1 | - | 2 | 157 | ||
AK015916.1 | - | 2 | 167 | |||
BC147640.1 | - | 2 | 145 | 9056630 | ||
BC147646.1 | - | 2 | 145 | 9056636 | ||
BC172114.1 | - | 2 | 145 | 9054400 |
