siRNA Details
Gene Symbol: Gga2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028758.2 | NP_083034.1 | 5 | 531 | ||
GenBank mRNA | AK004632.1 | - | 5 | 503 | ||
AK034612.1 | - | 2 | 174 | |||
AK129280.1 | - | 5 | 490 | |||
AK143691.1 | - | 5 | 519 | |||
AK148205.1 | - | 514 | ||||
AK149798.1 | - | 5 | 521 | |||
AK154904.1 | - | 5 | 506 | |||
AK159953.1 | - | 5 | 521 | |||
BC062933.1 | - | 5 | 500 | 30356060 |
