siRNA Details
Gene Symbol: Gga2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028758.2 | NP_083034.1 | 15 | 1668 | ||
GenBank mRNA | AK004632.1 | - | 15 | 1640 | ||
AK034612.1 | - | 12 | 1311 | |||
AK129280.1 | - | 15 | 1627 | |||
AK143691.1 | - | 15 | 1656 | |||
AK149798.1 | - | 15 | 1658 | |||
AK154904.1 | - | 15 | 1643 | |||
AK159953.1 | - | 15 | 1658 | |||
AK178573.1 | - | 3 | 381 | |||
AK183966.1 | - | 1 | 54 | |||
AK211643.1 | - | 1 | 54 | |||
AK211688.1 | - | 1 | 54 | |||
BC057377.1 | - | 8 | 1171 | 5698559 | ||
BC062933.1 | - | 15 | 1637 | 30356060 |
