siRNA Details
Gene Symbol: Armc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028768.3 | NP_083044.2 | 14 | 1532 | ||
GenBank mRNA | AK004793.2 | - | 14 | 1533 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Armc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028768.3 | NP_083044.2 | 14 | 1532 | ||
GenBank mRNA | AK004793.2 | - | 14 | 1533 |