siRNA Details
Gene Symbol: Armc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166138.1 | NP_001159610.1 | 4 | 470 | ||
NM_028768.3 | NP_083044.2 | 4 | 470 | |||
GenBank mRNA | AK004793.2 | - | 4 | 471 | ||
AK078280.1 | - | 4 | 221 | |||
AK083094.1 | - | 3 | 162 | |||
BC038866.1 | - | 4 | 428 | 1230321 |
