siRNA Details
Gene Symbol: Smg8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024262.1 | NP_077224.1 | 3 | 2132 | ||
GenBank mRNA | AK004714.1 | - | 3 | 2125 | ||
AK031106.1 | - | 3 | 2106 | |||
AK035089.1 | - | 3 | 1099 | |||
BC020005.1 | - | 3 | 2132 | 3981459 |
