siRNA Details
Gene Symbol: Josd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028792.3 | NP_083068.1 | 2, 3 | 1280 | ||
GenBank mRNA | AK004913.1 | - | 2, 3 | 1229 | ||
AK154799.1 | - | 2, 3 | 1329 | |||
AK214040.1 | - | 1, 2 | 144 | |||
BC006928.1 | - | 2, 3 | 428 | 2655826 | ||
BC086769.1 | - | 2, 3 | 400 | 5707990 |
