siRNA Details
Gene Symbol: Josd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028792.3 | NP_083068.1 | 3, 4 | 1456 | ||
GenBank mRNA | AK004913.1 | - | 3, 4 | 1405 | ||
AK154799.1 | - | 3, 4 | 1505 | |||
AK178708.1 | - | 1, 2 | 151 | |||
AK190593.1 | - | 1, 2 | 184 | |||
AK192247.1 | - | 1, 2 | 184 | |||
AK196858.1 | - | 1, 2 | 184 | |||
AK197352.1 | - | 1, 2 | 184 | |||
AK197502.1 | - | 1, 2 | 184 | |||
AK197953.1 | - | 1, 2 | 184 | |||
AK198562.1 | - | 1, 2 | 184 | |||
AK201245.1 | - | 1, 2 | 185 | |||
AK202351.1 | - | 1, 2 | 185 | |||
AK202639.1 | - | 1, 2 | 184 | |||
AK204805.1 | - | 356 | ||||
AK206589.1 | - | 1, 2 | 184 | |||
AK206648.1 | - | 1, 2 | 184 | |||
AK208563.1 | - | 1, 2 | 184 | |||
AK214040.1 | - | 2, 3 | 320 | |||
AK218856.1 | - | 1, 2 | 184 | |||
BC006928.1 | - | 3, 4 | 604 | 2655826 | ||
BC086769.1 | - | 3, 4 | 576 | 5707990 |

More Information
Other Species Targeted

Gene Ontology Categories:
