siRNA Details
Gene Symbol: 1700017B05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028820.2 | NP_083096.1 | 2 | 2087 | ||
GenBank mRNA | AK029230.1 | - | 2091 | |||
AK031730.1 | - | 2 | 2090 | |||
AK044396.1 | - | 2 | 2094 | |||
AK080676.1 | - | 2 | 2100 | |||
AK145074.1 | - | 2 | 2064 | |||
AK155015.1 | - | 2 | 2064 | |||
AK155487.1 | - | 2 | 2088 | |||
AK165963.1 | - | 2 | 2088 | |||
AK169617.1 | - | 2 | 2090 | |||
BC118527.1 | - | 2 | 1941 | 40098810 | ||
BC150963.1 | - | 2 | 1932 | 9087874 | ||
BC150965.1 | - | 2 | 1932 | 9087876 |
