siRNA Details
Gene Symbol: Mtmr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028860.2 | NP_083136.2 | 16 | 2525 | ||
GenBank mRNA | AK122261.1 | - | 1 | 145 | ||
BC026586.1 | - | 1 | 317 | |||
BC032166.1 | - | 16 | 2508 | 5352919 | ||
BC081544.1 | - | 16 | 2509 | 3672099 | ||
CB249296.1 | - | 1 | 169 |
