siRNA Details
Gene Symbol: Mgme1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289630.1 | NP_001276559.1 | 2 | 875 | ||
NM_001289631.1 | NP_001276560.1 | 2 | 546 | |||
NM_028984.5 | NP_083260.1 | 2 | 604 | |||
GenBank mRNA | AK018387.1 | - | 2 | 533 | ||
AK031535.1 | - | 2 | 474 | |||
AK043140.1 | - | 2 | 543 | |||
AK049565.1 | - | 2 | 745 | |||
AK159286.1 | - | 2 | 526 | |||
AK167919.1 | - | 2 | 533 | |||
BC063088.1 | - | 2 | 519 | 6329248 |
