siRNA Details
Gene Symbol: Mgme1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289630.1 | NP_001276559.1 | 2 | 541 | ||
NM_001289631.1 | NP_001276560.1 | 2 | 212 | |||
NM_028984.5 | NP_083260.1 | 2 | 270 | |||
GenBank mRNA | AK018387.1 | - | 2 | 199 | ||
AK031535.1 | - | 2 | 140 | |||
AK043140.1 | - | 2 | 209 | |||
AK049565.1 | - | 2 | 411 | |||
AK159286.1 | - | 2 | 192 | |||
AK167919.1 | - | 2 | 199 | |||
AK187747.1 | - | 1 | 115 | |||
AK188488.1 | - | 1 | 116 | |||
BC063088.1 | - | 2 | 185 | 6329248 | ||
BY136513.1 | - | 2 | 259 |
