siRNA Details
Gene Symbol: 4930451G09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271586.1 | NP_001258515.1 | 4 | 681 | ||
GenBank mRNA | AK015442.1 | - | 4 | 681 | ||
AK078576.1 | - | 1 | 55 | |||
AK154225.1 | - | 4 | 601 |
