siRNA Details
Gene Symbol: Atp13a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK156104.1 | - | 16 | 1721 | ||
AK171305.1 | - | 16 | 1695 | |||
BC023746.1 | - | 16 | 1717 | 5353766 | ||
BC042661.1 | - | 16 | 1687 | 4192094 |
