siRNA Details
Gene Symbol: Rprd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081293.1 | NP_001074762.1 | 11 | 3000 | ||
GenBank mRNA | AK083674.1 | - | 11 | 2769 | ||
AK086841.1 | - | 1 | 695 | |||
AK129150.1 | - | 10 | 2547 | |||
BC067054.1 | - | 11 | 2805 | 30542928 |
