siRNA Details
Gene Symbol: Ankrd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167757.1 | NP_001161229.1 | 1 | 227 | ||
NM_029202.2 | NP_083478.2 | 1 | 227 | |||
GenBank mRNA | AK015948.1 | - | 1 | 171 | ||
AK077080.1 | - | 1 | 196 | |||
BC049731.1 | - | 1 | 227 | 6771078 |
