siRNA Details
Gene Symbol: 1700020D05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023781.5 | NP_076270.3 | 1 | 849 | ||
GenBank mRNA | AF319980.1 | - | 1 | 813 | ||
AK006150.1 | - | 1 | 850 | |||
BC049585.1 | - | 1 | 846 | 6772543 | ||
BC061091.1 | - | 1 | 847 |
