siRNA Details
Gene Symbol: 1700026D08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029335.3 | NP_083611.2 | 2, 3 | 195 | ||
GenBank mRNA | AK006375.1 | - | 2, 3 | 183 | ||
BC048079.1 | - | 2, 3 | 175 | |||
BC061016.1 | - | 2, 3 | 189 | 6771412 |
