siRNA Details
Gene Symbol: Fam35a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029389.2 | NP_083665.2 | 4 | 749 | ||
GenBank mRNA | AK013957.1 | - | 3 | 674 | ||
AK149442.1 | - | 4 | 613 | |||
AK162134.1 | - | 2 | 560 | |||
BC144822.1 | - | 1 | 257 |
