siRNA Details
Gene Symbol: Smurf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038627.1 | NP_001033716.1 | 15 | 1998 | ||
NM_029438.3 | NP_083714.3 | 15 | 1998 | |||
GenBank mRNA | AK015264.1 | - | 11 | 1397 | ||
AK034736.1 | - | 6 | 596 | |||
AK122534.1 | - | 4 | 373 | |||
AK154491.1 | - | 15 | 1999 | |||
BC029097.1 | - | 15 | 2026 | 3660965 |
