siRNA Details
Gene Symbol: Smurf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038627.1 | NP_001033716.1 | 11 | 1448 | ||
NM_029438.3 | NP_083714.3 | 11 | 1448 | |||
GenBank mRNA | AK015264.1 | - | 7 | 847 | ||
AK034736.1 | - | 2 | 46 | |||
AK154491.1 | - | 11 | 1449 | |||
BC029097.1 | - | 11 | 1476 | 3660965 | ||
CB524571.1 | - | 5 | 526 | |||
CB527254.1 | - | 3 | 372 |
