siRNA Details
Gene Symbol: Vmp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029478.3 | NP_083754.2 | 10 | 1070 | ||
GenBank mRNA | AB047550.1 | - | 10 | 1082 | ||
AB076609.1 | - | 9 | 961 | |||
AK043091.1 | - | 10 | 1071 | |||
AK049559.1 | - | 10 | 1063 | |||
AK077443.1 | - | 10 | 1070 | |||
AK159468.1 | - | 10 | 1085 | |||
AK167173.1 | - | 10 | 1078 | |||
AK167202.1 | - | 10 | 1078 | |||
AK211254.1 | - | 2 | 132 | |||
AK219288.1 | - | 2 | 132 | |||
AX775963.1 | - | 10 | 1040 | |||
BC004013.1 | - | 10 | 1064 | 3493738 |
