siRNA Details
Gene Symbol: Mgea5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023799.3 | NP_076288.1 | 7 | 1038 | ||
GenBank mRNA | AF132214.1 | - | 7 | 883 | ||
AK017997.1 | - | 1 | 116 | |||
AK088774.1 | - | 7 | 1301 | |||
AK129188.1 | - | 7 | 1267 | |||
AK145227.1 | - | 7 | 1288 | |||
BC020093.1 | - | 7 | 975 | |||
BC041109.1 | - | 7 | 1368 | |||
BC054821.1 | - | 7 | 1038 | 6515250 |
Gene Symbol: D19Ertd409e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK163010.1 | - | 1 | 1719 | ||
BG080263.2 | - | 1 | 196 |
