siRNA Details
Gene Symbol: Mgea5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023799.3 | NP_076288.1 | 9 | 1417 | ||
GenBank mRNA | AF132214.1 | - | 9 | 1262 | ||
AK017997.1 | - | 3 | 495 | |||
AK088774.1 | - | 9 | 1680 | |||
AK129188.1 | - | 9 | 1646 | |||
AK145227.1 | - | 9 | 1667 | |||
BC020093.1 | - | 9 | 1354 | |||
BC041109.1 | - | 9 | 1747 | |||
BC054821.1 | - | 9 | 1417 | 6515250 |
