siRNA Details
Gene Symbol: Mgea5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023799.3 | NP_076288.1 | 9 | 1910 | ||
GenBank mRNA | AF132214.1 | - | 9 | 1755 | ||
AK129188.1 | - | 9 | 2139 | |||
AK145227.1 | - | 9 | 2160 | |||
BC020093.1 | - | 9 | 1847 | |||
BC041109.1 | - | 9 | 2240 | |||
BC054821.1 | - | 9 | 1910 | 6515250 |
