siRNA Details
Gene Symbol: Gpsm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029522.2 | NP_083798.2 | 7 | 1282 | ||
GenBank mRNA | AK040996.1 | - | 7 | 1171 | ||
AK143552.1 | - | 7 | 1283 | |||
AY081187.1 | - | 7 | 1126 | |||
BC021308.1 | - | 6 | 837 | 5007832 |
