siRNA Details
Gene Symbol: Gpsm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029522.2 | NP_083798.2 | 15 | 2416 | ||
GenBank mRNA | AK018084.1 | - | 5 | 663 | ||
AK143552.1 | - | 15 | 2417 | |||
AY081187.1 | - | 15 | 2260 | |||
BC021308.1 | - | 14 | 1971 | 5007832 |
