siRNA Details
Gene Symbol: Ccdc138 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162956.1 | NP_001156428.1 | 4, 5 | 360 | ||
GenBank mRNA | BC119028.1 | - | 4, 5 | 349 | 8733777 | |
BC119030.1 | - | 4, 5 | 349 | 8733779 |
