siRNA Details
Gene Symbol: Ankrd36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023816.2 | NP_076305.2 | 4 | 818 | ||
GenBank mRNA | AF294328.1 | - | 4 | 820 | ||
AK005925.1 | - | 4 | 792 | |||
AK132751.1 | - | 4 | 819 | |||
AK134466.1 | - | 1 | 228 | |||
BC120815.1 | - | 4 | 596 | |||
BC120817.1 | - | 4 | 593 |
