siRNA Details
Gene Symbol: 1700016D06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024271.1 | NP_077233.1 | 5 | 489 | ||
GenBank mRNA | AK006019.1 | - | 5 | 462 | ||
AY026045.1 | - | 5 | 489 | |||
BC048664.1 | - | 5 | 481 | 6773850 |
