siRNA Details
Gene Symbol: Snx7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190156.1 | NP_001177085.1 | 7 | 1104 | ||
NM_029655.3 | NP_083931.2 | 7 | 1104 | |||
GenBank mRNA | AK011015.1 | - | 7 | 1079 | ||
AK038971.1 | - | 7 | 1105 | |||
AK043893.1 | - | 1 | 769 | |||
AK052487.1 | - | 7 | 1061 | |||
AK163631.1 | - | 1 | 1666 | |||
AK170966.1 | - | 7 | 1051 | |||
BC087942.1 | - | 7 | 962 | 30303462 |
