siRNA Details
Gene Symbol: Immt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253681.1 | NP_001240610.1 | 15 | 2330 | ||
NM_001253686.1 | NP_001240615.1 | 14 | 2315 | |||
NM_001253687.1 | NP_001240616.1 | 13 | 2219 | |||
NM_001253688.1 | NP_001240617.1 | 13 | 2129 | |||
NM_001253689.1 | NP_001240618.1 | 13 | 2100 | |||
NM_029673.3 | NP_083949.2 | 15 | 2363 | |||
GenBank mRNA | AK006977.1 | - | 3 | 708 | ||
AK033126.1 | - | 13 | 2130 | |||
AK038129.1 | - | 15 | 2353 | |||
AK049189.1 | - | 15 | 2274 | |||
AK146640.1 | - | 13 | 2092 | |||
AK152588.1 | - | 14 | 2261 | |||
AK159909.1 | - | 14 | 2266 | |||
AK169365.1 | - | 13 | 2032 | |||
AK206799.1 | - | 1 | 178 | |||
BC025034.1 | - | 3 | 1109 | |||
BC055840.1 | - | 3 | 792 | |||
BC061010.1 | - | 4 | 834 | |||
BC080790.1 | - | 2 | 680 | |||
BC108356.1 | - | 7 | 1261 | |||
BC145704.1 | - | 2 | 643 | |||
BC145706.1 | - | 2 | 643 | |||
BU754623.1 | - | 1 | 411 |
