siRNA Details
Gene Symbol: Mospd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290523.1 | NP_001277452.1 | 8 | 773 | ||
NM_001290524.1 | NP_001277453.1 | 8 | 773 | |||
NM_029730.4 | NP_084006.2 | 8 | 773 | |||
GenBank mRNA | AK010480.1 | - | 8 | 695 | ||
AK031405.1 | - | 8 | 698 | |||
AK035985.1 | - | 8 | 733 | |||
BC026425.1 | - | 8 | 699 | 4223175 |
