siRNA Details
Gene Symbol: Rnft1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029788.5 | NP_084064.1 | 4, 5 | 757 | ||
GenBank mRNA | AK002624.1 | - | 4, 5 | 715 | ||
AK036401.1 | - | 4, 5 | 758 | |||
AK153293.1 | - | 4, 5 | 726 | |||
AK171077.1 | - | 3, 4 | 2008 | |||
BC037057.1 | - | 3, 4 | 544 | |||
BC050796.1 | - | 4, 5 | 725 | 6743413 |
