siRNA Details
Gene Symbol: Hnrnpm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109913.1 | NP_001103383.1 | 15 | 1922 | ||
NM_029804.3 | NP_084080.1 | 14 | 2039 | |||
GenBank mRNA | AK011521.1 | - | 14 | 1985 | ||
AK133911.1 | - | 13 | 1921 | |||
AK145343.1 | - | 15 | 1860 | |||
AK159848.1 | - | 15 | 1854 | |||
AK163866.1 | - | 3 | 871 | |||
AK166670.1 | - | 14 | 1974 | |||
AK168347.1 | - | 15 | 1857 | |||
AK169101.1 | - | 15 | 1856 | |||
AK171892.1 | - | 15 | 1857 | |||
AK178014.1 | - | 1 | 47 | |||
AK219463.1 | - | 1 | 47 | |||
AK220299.1 | - | 15 | 1882 | |||
BC005758.1 | - | 3 | 932 | |||
BC017539.1 | - | 15 | 1852 | |||
BC035525.1 | - | 15 | 1857 | |||
BC054785.1 | - | 3441 | ||||
BC065172.1 | - | 15 | 1852 | 6415170 |
