siRNA Details
Gene Symbol: Cep83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029852.2 | NP_084128.2 | 5 | 808 | ||
GenBank mRNA | AK015007.1 | - | 5 | 945 | ||
AK141627.1 | - | 5 | 929 | |||
BC028960.1 | - | 4 | 509 | |||
BC070464.1 | - | 5 | 808 | 30608497 |
