siRNA Details
Gene Symbol: Hook1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030014.2 | NP_084290.1 | 5 | 443 | ||
GenBank mRNA | AF487912.1 | - | 5 | 525 | ||
AK020924.1 | - | 5 | 467 | |||
AK038941.1 | - | 5 | 454 | |||
AK043870.1 | - | 5 | 450 | |||
AK049897.1 | - | 5 | 444 | |||
BC061688.1 | - | 5 | 443 | 5364426 |
