siRNA Details
Gene Symbol: Dnajc21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030046.2 | NP_084322.2 | 12 | 1629 | ||
GenBank mRNA | AK020688.1 | - | 3 | 534 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dnajc21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030046.2 | NP_084322.2 | 12 | 1629 | ||
GenBank mRNA | AK020688.1 | - | 3 | 534 |