siRNA Details
Gene Symbol: Zfp687 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030074.2 | NP_084350.1 | 3 | 2403 | ||
GenBank mRNA | AK019851.2 | - | 3 | 2404 | ||
AK129360.1 | - | 2 | 1606 | |||
AK133173.1 | - | 3 | 2390 | |||
AK148239.1 | - | 3 | 2360 | |||
BC056943.1 | - | 3 | 2393 | 6831344 | ||
BC060179.1 | - | 3 | 2393 | 6831344 |

More Information
Other Species Targeted

Gene Ontology Categories:
