siRNA Details
Gene Symbol: Zfp687 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030074.2 | NP_084350.1 | 2 | 1760 | ||
GenBank mRNA | AK019851.2 | - | 2 | 1761 | ||
AK129360.1 | - | 1 | 963 | |||
AK133173.1 | - | 2 | 1747 | |||
AK148239.1 | - | 2 | 1717 | |||
BC056943.1 | - | 2 | 1750 | 6831344 | ||
BC060179.1 | - | 2 | 1750 | 6831344 |
