siRNA Details
Gene Symbol: Nav2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111016.1 | NP_001104486.1 | 8 | 1507 | ||
NM_175272.3 | NP_780481.2 | 7 | 1769 | |||
GenBank mRNA | AK040152.1 | - | 6 | 1054 | ||
AK084063.1 | - | 8 | 1485 | |||
AK129480.1 | - | 7 | 1064 | |||
BC038177.1 | - | 2 | 198 |
