siRNA Details
Gene Symbol: Nav2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111016.1 | NP_001104486.1 | 7 | 1351 | ||
NM_175272.3 | NP_780481.2 | 6 | 1613 | |||
GenBank mRNA | AK031334.1 | - | 6 | 911 | ||
AK040152.1 | - | 5 | 898 | |||
AK084063.1 | - | 7 | 1329 | |||
AK129480.1 | - | 6 | 908 | |||
BC150679.1 | - | 5 | 601 |
