siRNA Details
Gene Symbol: Brd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289606.1 | NP_001276535.1 | 18 | 2648 | ||
NM_001289607.1 | NP_001276536.1 | 18 | 2657 | |||
NM_030147.3 | NP_084423.2 | 19 | 2867 | |||
GenBank mRNA | AK032320.1 | - | 19 | 2579 | ||
AK132365.1 | - | 19 | 2563 | |||
AK141387.1 | - | 3 | 234 | |||
AK161689.1 | - | 18 | 2361 | |||
BC021482.1 | - | 17 | 3696 | |||
BC023160.1 | - | 4 | 480 | |||
BC023837.1 | - | 18 | 2373 | |||
BC025644.1 | - | 19 | 2612 | 5323755 |
