siRNA Details
Gene Symbol: Brd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289606.1 | NP_001276535.1 | 16 | 2468 | ||
NM_001289607.1 | NP_001276536.1 | 16 | 2477 | |||
NM_030147.3 | NP_084423.2 | 17 | 2687 | |||
GenBank mRNA | AK032320.1 | - | 17 | 2399 | ||
AK132365.1 | - | 17 | 2383 | |||
AK141387.1 | - | 1 | 54 | |||
AK161689.1 | - | 16 | 2181 | |||
BC021482.1 | - | 15 | 3516 | |||
BC023160.1 | - | 2 | 300 | |||
BC023837.1 | - | 16 | 2193 | |||
BC025644.1 | - | 17 | 2432 | 5323755 |
