siRNA Details
Gene Symbol: Mctp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030174.2 | NP_084450.2 | 7 | 846 | ||
GenBank mRNA | AK047562.1 | - | 7 | 847 | ||
AK054478.1 | - | 3 | 219 | |||
AK154322.1 | - | 6 | 661 |
