siRNA Details
Gene Symbol: Brpf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282126.1 | NP_001269055.1 | 5, 6 | 2297 | ||
NM_001282127.1 | NP_001269056.1 | 5, 6 | 2065 | |||
NM_001282128.1 | NP_001269057.1 | 5, 6 | 2297 | |||
NM_030178.2 | NP_084454.1 | 5, 6 | 2065 | |||
GenBank mRNA | AK043372.1 | - | 5, 6 | 2298 | ||
AK156375.1 | - | 5, 6 | 2245 | |||
BC005647.1 | - | 3, 4 | 337 | |||
BC031594.1 | - | 3, 4 | 971 | |||
BC046521.1 | - | 5, 6 | 2005 | 5363697 | ||
BC138361.1 | - | 5, 6 | 1889 | 8861381 | ||
BC145256.1 | - | 5, 6 | 2121 | 9053809 |
