siRNA Details
Gene Symbol: Brpf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282126.1 | NP_001269055.1 | 3 | 1200 | ||
NM_001282127.1 | NP_001269056.1 | 3 | 968 | |||
NM_001282128.1 | NP_001269057.1 | 3 | 1200 | |||
NM_030178.2 | NP_084454.1 | 3 | 968 | |||
GenBank mRNA | AK043372.1 | - | 3 | 1201 | ||
AK156375.1 | - | 3 | 1148 | |||
AK187717.1 | - | 1 | 67 | |||
AK199235.1 | - | 1 | 67 | |||
AK201246.1 | - | 1 | 67 | |||
AK203431.1 | - | 1 | 67 | |||
BC046521.1 | - | 3 | 908 | 5363697 | ||
BC138361.1 | - | 3 | 792 | 8861381 | ||
BC145256.1 | - | 3 | 1024 | 9053809 | ||
DQ288860.1 | - | 2 | 757 |
