siRNA Details
Gene Symbol: Gatsl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030719.3 | NP_109644.2 | 3 | 579 | ||
GenBank mRNA | AJ296173.1 | - | 3 | 619 | ||
AK039122.1 | - | 3 | 605 | |||
AK052217.1 | - | 3 | 564 | |||
BC094613.1 | - | 3 | 579 | 6856325 |
